missing translation for 'onlineSavingsMsg'
Learn More

Aminoguanidine Hydrochloride, TRC

Product Code. 17992885
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17992885 1 g 1g
17904835 5 g 5g
17984155 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17992885 Supplier Toronto Research Chemicals Supplier No. TRCA6098631G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1937-19-5
Molecular Formula CH6N4 . HCl
Molecular Weight (g/mol) 110.55
Synonym Amino-guanidine Monohydrochloride (8CI), Hydrazinecarboximidamide, Monohydrochloride (9CI), Aminoguanidine Monohydrochloride, Aminoguanidinium Chloride, GER 11, Pimagedine Hydrochloride
IUPAC Name 2-aminoguanidine;hydrochloride
SMILES Cl.NNC(=N)N

Specifications

Quantity 1 g
InChI Formula InChI=1S/CH6N4.ClH/c2-1(3)5-4;/h4H2,(H4,2,3,5);1H
Chemical Name or Material Aminoguanidine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.