missing translation for 'onlineSavingsMsg'
Learn More

Aminopyralid, TRC

Product Code. 30133897
Change view
Click to view available options
Quantity:
1 mg
5 mg
50 mg
Unit Size:
1mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133897 50 mg 50mg
30132785 1 mg 1mg
17950905 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133897 Supplier Toronto Research Chemicals Supplier No. TRCA62882050MG
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150114-71-9
Molecular Formula C6 H4 Cl2 N2 O2
Molecular Weight (g/mol) 207.01
Synonym 4-Amino-3,6-dichloro-2-pyridinecarboxylic acid, 4-Amino-3,6-dichloropicolinic acid, 4-Amino-3,6-dichloropyridine-2-carboxylic acid, Aminopyralid, Milestone, Milestone (aminopyralid herbicide)
IUPAC Name 4-amino-3,6-dichloropyridine-2-carboxylic acid
SMILES Nc1cc(Cl)nc(C(=O)O)c1Cl

Specifications

Analyte or Component Names Aminopyralid
Shipping Condition Room Temperature
InChI Formula InChI=1S/C6H4Cl2N2O2/c7-3-1-2(9)4(8)5(10-3)6(11)12/h1H,(H2,9,10)(H,11,12)
Solution Type Neat
Industry Type Pharmaceutical
Quantity 50 mg
Formula Weight 205.965
Percent Purity >95%
Chemical Name or Material Aminopyralid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.