missing translation for 'onlineSavingsMsg'
Learn More

Amiodarone hydrochloride, MedChemExpress

Product Code. 16488607
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488607 5 g 5g
16461399 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16488607 Supplier Medchem Express Supplier No. HY14188
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Amiodarone hydrochloride, a benzofuran-based Class III antiarrhythmic agent, inhibits WT outwardIhERG tails with an IC50 of ∼45 nM. Amiodarone hydrochloride induces cell proliferation and myofibroblast differentiation via ERK1/2 and p38 MAPK signaling in fibroblasts. Amiodarone hydrochloride can be used in the research of both supraventricular and ventricular arrhythmias.

Amiodarone hydrochloride, a benzofuran-based Class III antiarrhythmic agent, inhibits WT outwardIhERG tails with an IC50 of ∼45 nM. Amiodarone hydrochloride induces cell proliferation and myofibroblast differentiation via ERK1/2 and p38 MAPK signaling in fibroblasts. Amiodarone hydrochloride can be used in the research of both supraventricular and ventricular arrhythmias.

Specifications

Chemical Name or Material Amiodarone hydrochloride
CAS 19774-82-4
Color White
Purity Grade Notes Research
Molecular Formula C25H30ClI2NO3
Quantity 5 g
Solubility Information DMSO : 24.5 mg/mL (35.94 mM; Need ultrasonic and warming)
SMILES CCCCC1=C(C(C2=CC(I)=C(OCCN(CC)CC)C(I)=C2)=O)C3=C(O1)C=CC=C3.Cl
Molecular Weight (g/mol) 681.77
Formula Weight 681.77
Percent Purity 99.67%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.