missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Amisulpride

Product Code. 15731809 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15731809 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15731809 Supplier Thermo Scientific Chemicals Supplier No. 462300010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 71675-85-9 | C17H27N3O4S | 369.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 71675-85-9
Melting Point 127.0°C
Molecular Formula C17H27N3O4S
Quantity 1 g
Synonym amisulpride, aminosultopride, solian, amisulprida, amisulpridum, deniban, amisulpridum inn-latin, amisulprida inn-spanish, amisulpride inn, amisulpiride
InChI Key NTJOBXMMWNYJFB-UHFFFAOYSA-N
SMILES CCN1CCCC1CNC(=O)C2=CC(=C(C=C2OC)N)S(=O)(=O)CC
IUPAC Name 4-amino-N-[(1-ethylpyrrolidin-2-yl)methyl]-5-ethylsulfonyl-2-methoxybenzamide
Molecular Weight (g/mol) 369.48
PubChem CID 2159
ChEBI CHEBI:64045
Formula Weight 369.48
Chemical Name or Material Amisulpride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.