missing translation for 'onlineSavingsMsg'
Learn More

Amitriptyline hydrochloride, MedChemExpress

Product Code. 16450978
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16450978 5 g 5g
16432219 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16450978 Supplier Medchem Express Supplier No. HYB0527A
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Amitriptyline hydrochloride is an inhibitor of serotonin reuptake transporter (SERT) and noradrenaline reuptake transporter (NET), with Kis of 3.45 nM and 13.3 nM for human SERT and NET, respectively.

Amitriptyline hydrochloride also weakly binds to dopamine reuptake transporter (DAT) with a Ki of 2.58 μM. Amitriptyline hydrochloride also inhibits adrenergic, muscarinic, histamine and 5-HT receptors. Amitriptyline hydrochloride is a TrkA and TrkB receptors agonist with potent neurotrophic activity. Amitriptyline hydrochloride has antidepressant activity.

Specifications

Chemical Name or Material Amitriptyline hydrochloride
Physical Form Solid
CAS 549-18-8
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C20H24ClN
For Use With (Application) Neuroscience-Neurodegeneration
Solubility Information DMSO : ≥ 100 mg/mL (318.61 mM) ∣H2O : ≥ 50 mg/mL (159.31 mM)
SMILES CN(C)CC/C=C1C2=CC=CC=C2CCC3=C\1C=CC=C3.Cl
Molecular Weight (g/mol) 313.86
Formula Weight 313.86
Percent Purity 99.56%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.