missing translation for 'onlineSavingsMsg'
Learn More

Amitriptyline Hydrochloride, TRC

Product Code. 30136257
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136257 1 g 1g
30132896 10 g 10g
18042910 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136257 Supplier Toronto Research Chemicals Supplier No. TRCA6333501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 549-18-8
Formula Weight 313.1597
InChI Formula InChI=1S/C20H23N.ClH/c1-21(2)15-7-12-20-18-10-5-3-8-16(18)13-14-17-9-4-6-11-19(17)20;/h3-6,8-12H,7,13-15H2,1-2H3;1H
IUPAC Name 3-(5,6-dihydrodibenzo[2,1-b:1',2'-e][7]annulen-11-ylidene)-N,N-dimethylpropan-1-amine;hydrochloride
Molecular Formula C20 H23 N . Cl H
Molecular Weight (g/mol) 313.86
Percent Purity >95%
SMILES Cl.CN(C)CCC=C1c2ccccc2CCc3ccccc13
Synonym Amitriptyline Hydrochloride, Nortriptyline Hydrochloride Imp. F (EP) as Hydrochloride, 1-Propanamine, 3-(10,11-dihydro-5H-dibenzo[a,d]cyclohepten-5-ylidene)-N,N-dimethyl-, hydrochloride (1:1), 1-Propanamine, 3-(10,11-dihydro-5H-dibenzo[a,d]cyclohepten-5-ylidene)-N,N-dimethyl-, hydrochloride (9CI), 5H-Dibenzo[a,d]cycloheptene-δ5,γ-propylamine, 10,11-dihydro-N,N-dimethyl-, hydrochloride (6CI,8CI), 10,11-Dihydro-N,N-dimethyl-5H-dibenzo[a,d]cycloheptene-δ5,γ-propylamine hydrochloride, 5-(3-Dimethylaminopropylidene)dibenzo[a,d][1,4]cycloheptadiene hydrochloride, ADT, Adepril, Amicen, Amil 25, Amilent, Amilit, Amineurin, Amiplin, Amiprin, Amitid, Amitril, Amitrip, Amitriptyline chloride, Amyline, Amyzol, Anapsique, Apo-Amitriptyline, Damilen hydrochloride, Deprex, Domical, Elatrol, Elatrolet, Elavil, Enafon, Endep, Etravil, Euplit, Lantron, Laroxyl, Larozyl, Lentizol, Miketorin, NIH 10794, Noriline, Novoprotect, Pinsanu, Pinsaun, Redomex, Ro 4-1575, Saroten, Saroten Retard, Sarotena, Sarotex, Sylvemid, Syneudon, Teperin, Trepiline, Tridep, Tripta, Triptizol, Trynol, Tryptal, Tryptanol, Tryptine, Tryptizol, Trytomer, Uxen, Vanatrip
Chemical Name or Material Amitriptyline Hydrochloride
Grade HPLC
Show More Show Less

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.