missing translation for 'onlineSavingsMsg'
Learn More

Amitriptyline N-Oxide, TRC

Product Code. 30004155
Change view
Click to view available options
Quantity:
10 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004155 100 mg 100mg
30009741 10 mg 10mg
30003947 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004155 Supplier Toronto Research Chemicals Supplier No. TRCA633355100MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4317-14-0
Molecular Formula C20 H23 N O
Molecular Weight (g/mol) 293.4
Synonym 3-(10,11-Dihydro-5H-dibenzo[a,d]cyclohepten-5-ylidene)-N,N-dimethyl-1-propanamine N-Oxide, 10,11-Dihydro-N,N-dimethyl-5H-Dibenzo[a,d]cycloheptene-δ5,γ-propylamine N-Oxide, Amitriptyline Oxide, Amitriptylinoxide, Equilibrin
IUPAC Name 3-(5,6-dihydrodibenzo[2,1-b:1',2'-e][7]annulen-11-ylidene)-N,N-dimethylpropan-1-amine oxide
SMILES C[N+](C)([O-])CCC=C1c2ccccc2CCc3ccccc13

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C20H23NO/c1-21(2,22)15-7-12-20-18-10-5-3-8-16(18)13-14-17-9-4-6-11-19(17)20/h3-6,8-12H,7,13-15H2,1-2H3
Formula Weight 293.178
Chemical Name or Material Amitriptyline N-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.