missing translation for 'onlineSavingsMsg'
Learn More

Amlexanox, MedChemExpress

Product Code. 16478426
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478426 100 mg 100mg
16464457 50 mg 50mg
16428588 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16478426 Supplier Medchem Express Supplier No. HYB0713
 Restricted Item
Estimated Shipment: 14-12-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

MedChemExpress Amlexanox (AA673; Amoxanox; CHX3673) is a specific inhibitor of IKKε and TBK1, and inhibits the IKKε and TBK1 activity determined by MBP phosphorylation with an IC50 of approximately 1-2 μM.

Amlexanox (AA673

Specifikationer

Chemical Name or Material Amlexanox
CAS 68302-57-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H14N2O4
Quantity 100 mg
Synonym AA673 Amoxanox CHX3673
Solubility Information DMSO : 100 mg/mL (335.24 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=C(N)N=C2C(C(C3=CC(C(C)C)=CC=C3O2)=O)=C1)O
Molecular Weight (g/mol) 298.29
Formula Weight 298.29
Percent Purity 99.73%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Visa mer Visa mindre

Research purposes only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.