Learn More
Invitrogen™ Amlitelimab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559062
 View more versions of this product
Product Code. 30282548
  • 1204.00 € / 1mg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Amlitelimab is a monoclonal antibody that targets and inhibits OX40L, a protein involved in the activation and survival of T cells, thereby modulating the immune response.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Amlitelimab
Recombinant Monoclonal
Unconjugated
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM Histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Protein A
RUO
Human
Antibody
IgG4SP