missing translation for 'onlineSavingsMsg'
Learn More

Ammonium cobalt(II) sulfate hexahydrate, 99%

Product Code. 11304948
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11304948

Brand: Thermo Scientific Alfa Aesar 043902.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

As spectroscopic standard, cobalt plating, catalysis, ceramics | CAS: 13586-38-4 | H8CoN2O8S2·6H2O

Ammonium cobalt(II) sulfate hexahydrate is uses as spectroscopic standard, cobalt plating, catalysis and ceramics. It is also used in pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13586-38-4
Molecular Formula H8CoN2O8S2·6H2O
MDL Number MFCD00151049
Synonym Cobalt(II) ammonium sulfate hexhydrate; Cobaltous ammonium sulfate hexahydrate

Specifications

Melting Point ∼120°C (decomposition)
Color Pink to Red
Physical Form Crystalline
Quantity 100 g
Linear Formula (NH4)2Co(SO4)2·6H2O
UN Number UN3077
Solubility Information Soluble in water, Insoluble in alcohol
Formula Weight 395.22 (287.14 Anhydrous)
Percent Purity ≥99%
Odor Odorless
Sensitivity Hygroscopic
Density 1.902 g/mL
Chemical Name or Material Ammonium cobalt (II) sulfate hexahydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.