missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium fluoride - hydrofluoric acid mixture, Solstice
Shop All Honeywell Research Chemicals ProductsDescription
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications
Specifications
| CAS | 1341-49-7, 12125-01-8, 7732-18-5 |
| Boiling Point | 100°C |
| CAS Min % | ≥10.00%, ≥20.00%, ≥50.00% |
| CAS Max % | <20.00%, <30.00%, <70.00% |
| Purity Grade Notes | Semiconductor grade reagent |
| Packaging | Plastic Bottle |
| Quantity | 2.5 L |
| Physical Form | Liquid, clear |
Safety and Handling
Product Identifier
- Ammonium fluoride - hydrofluoric acid mixture
- Danger
- Acute toxicity Category 4
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contain: ammonium fluoride, ammonium bifluoride
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?