missing translation for 'onlineSavingsMsg'
Learn More

Ammonium fluoride - hydrofluoric acid mixture, Solstice

Product Code. 16851200 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 L
Unit Size:
1 pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16851200 2.5 L 1 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16851200 Supplier Honeywell Research Chemicals Supplier No. 40207H2.5L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Semiconductor Grade, AF 875-125, etching mixture

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1341-49-7, 12125-01-8, 7732-18-5
Boiling Point 100°C
CAS Min % ≥10.00%, ≥20.00%, ≥50.00%
CAS Max % <20.00%, <30.00%, <70.00%
Purity Grade Notes Semiconductor grade reagent
Packaging Plastic Bottle
Quantity 2.5 L
Physical Form Liquid, clear
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.