missing translation for 'onlineSavingsMsg'
Learn More

Ammonium fluoride solution, Honeywell™

Product Code. 16606173 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
5 L
Unit Size:
5 L
Product Code. Quantity unitSize
16606173 5 L 5 L
1 options
This item is not returnable. View return policy

Product Code. 16606173

Brand: Honeywell Research Chemicals 40208H5L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Semiconductor Grade (Honeywell 17767)

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Specifications

CAS 12125-01-8, 7732-18-5
Boiling Point > 100°C (1.013 hPa)
CAS Min % ≥30.00%, ≥50.00%
CAS Max % <50.00%, <70.00%
Purity Grade Notes Semiconductor grade reagent
MDL Number MFCD00011423
Molecular Weight (g/mol) 37.04
Packaging Plastic Can
Quantity 5 L
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.