missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexafluorotitanate(IV), 99.99%, (trace metal basis)

Product Code. 10432201 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10432201 10 g Glass bottle 10g
10194160 50 g Glass bottle 50g
10677684 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10432201 Supplier Thermo Scientific Acros Supplier No. 194350100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16962-40-6 | H8F6N2Ti | 197.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16962-40-6
Molecular Formula H8F6N2Ti
Molecular Weight (g/mol) 197.97
MDL Number MFCD00010888

Specifications

Color White
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 10 g
Assay Percent Range 99.99%
Linear Formula (NH4)2TiF6
Solubility Information Solubility in water: insoluble
Formula Weight 197.97
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Ammonium hexafluorotitanate(IV)
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium hexafluorotitanate(IV)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.