missing translation for 'onlineSavingsMsg'
Learn More

Ammonium magnesium phosphate hexahydrate, 98%

Product Code. 11468107
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11468107 100 g 100g
11478107 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11468107 Supplier Thermo Scientific Alfa Aesar Supplier No. A17497.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 13478-16-5 | H16MgNO10P | 245.404 g/mol

It is used as pharmaceutical intermediae.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediae.

Solubility
Insoluble in water.

Notes
Store in a cool place. Ensure proper ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13478-16-5
Molecular Formula H16MgNO10P
Molecular Weight (g/mol) 245.404
MDL Number MFCD00150461
InChI Key CKMXBZGNNVIXHC-UHFFFAOYSA-L
Synonym unii-aw3ejl1462, magnesium ammonium phosphate hexahydrate, acmc-20alsk, magnesium 2+ ion ammonium hexahydrate phosphate, magnesium 2+ ammonium hexahydrate phosphate, 7785-21-9 h3po4-ammonium-magnesium 1:1:1
PubChem CID 10220511
IUPAC Name azanium;magnesium;phosphate;hexahydrate
SMILES [NH4+].O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Mg+2]

Specifications

Quantity 100 g
Solubility Information Insoluble in water.
Formula Weight 245.41 (137.32 Anhydrous)
Percent Purity 98%
Density 1.711
Chemical Name or Material Ammonium magnesium phosphate hexahydrate
compliance-icons
Product Identifier
  • Ammonium magnesium phosphate hexahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Ammonium magnesium phosphate hexahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.