missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium metavanadate, 99.996%, (trace metal basis)
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
Specifications
Specifications
| Melting Point | 200.0°C |
| Color | White to Yellow |
| Physical Form | Crystalline Powder |
| Quantity | 10 g |
| Assay Percent Range | 99.996% |
| Linear Formula | NH4VO3 |
| Fieser | 01,1057 |
| Merck Index | 15,565 |
| Solubility Information | Solubility in water: 5.1g/L (20°C). Other solubilities: insoluble in ethanol, 69.5g/L (96°C) |
| IUPAC Name | azanium;oxido(dioxo)vanadium |
| Show More |
Safety and Handling
Product Identifier
- Ammonium metavanadate
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
- H372-Causes damage to organs.
- H319-Causes serious eye irritation.
- H361d-Suspected of damaging the unborn child.
- H411-Toxic to aquatic life with long lasting effects.
- H332-Harmful if inhaled.
- H301-Toxic if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction