CAS RN 7803-55-6
Ammonium metavanadate, 99.5%, for analysis
CAS: 7803-55-6 Molecular Formula: H4NO3V Molecular Weight (g/mol): 116.98 MDL Number: MFCD00011430 InChI Key: YBVKNHXQSRDWAA-UHFFFAOYSA-M Synonym: ammonium metavanadate,ammonium vanadate v,ammonium monovanadate,unii-fl85px638g,vanadate vo31-, ammonium,vanadate vo31-, ammonium 1:1,ammonium vanadium oxide,ammoniummetavanadate,vanadic acid, hvo3 , ammonium salt,ammonium meta-vanadate PubChem CID: 516859 SMILES: N.O[V](=O)=O
Ammonium metavanadate, ACS reagent
CAS: 7803-55-6 Molecular Formula: H4NO3V Molecular Weight (g/mol): 116.98 MDL Number: MFCD00011430 InChI Key: YBVKNHXQSRDWAA-UHFFFAOYSA-M Synonym: ammonium metavanadate,ammonium vanadate v,ammonium monovanadate,unii-fl85px638g,vanadate vo31-, ammonium,vanadate vo31-, ammonium 1:1,ammonium vanadium oxide,ammoniummetavanadate,vanadic acid, hvo3 , ammonium salt,ammonium meta-vanadate PubChem CID: 516859 SMILES: N.O[V](=O)=O
Ammonium vanadium oxide, Puratronic™, 99.995% (metals basis)
CAS: 7803-55-6 Molecular Formula: H4NO3V Molecular Weight (g/mol): 116.98 MDL Number: MFCD00011430 InChI Key: YBVKNHXQSRDWAA-UHFFFAOYSA-M Synonym: ammonium metavanadate,ammonium vanadate v,ammonium monovanadate,unii-fl85px638g,vanadate vo31-, ammonium,vanadate vo31-, ammonium 1:1,ammonium vanadium oxide,ammoniummetavanadate,vanadic acid, hvo3 , ammonium salt,ammonium meta-vanadate PubChem CID: 516859 IUPAC Name: vanadiumoylol amine SMILES: N.O[V](=O)=O
Ammonium metavanadate, 99.996%, (trace metal basis)
CAS: 7803-55-6 Molecular Formula: H4NO3V Molecular Weight (g/mol): 116.98 MDL Number: MFCD00011430 InChI Key: YBVKNHXQSRDWAA-UHFFFAOYSA-M Synonym: ammonium metavanadate,ammonium vanadate v,ammonium monovanadate,unii-fl85px638g,vanadate vo31-, ammonium,vanadate vo31-, ammonium 1:1,ammonium vanadium oxide,ammoniummetavanadate,vanadic acid, hvo3 , ammonium salt,ammonium meta-vanadate PubChem CID: 516859 SMILES: N.O[V](=O)=O
Ammonium vanadium oxide, 99% (metals basis)
CAS: 7803-55-6 Molecular Formula: H4NO3V Molecular Weight (g/mol): 116.98 MDL Number: MFCD00011430 InChI Key: YBVKNHXQSRDWAA-UHFFFAOYSA-M Synonym: ammonium metavanadate,ammonium vanadate v,ammonium monovanadate,unii-fl85px638g,vanadate vo31-, ammonium,vanadate vo31-, ammonium 1:1,ammonium vanadium oxide,ammoniummetavanadate,vanadic acid, hvo3 , ammonium salt,ammonium meta-vanadate PubChem CID: 516859 SMILES: N.O[V](=O)=O
Vanadium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
CAS: 7803-55-6 Molecular Formula: H4NO3V Molecular Weight (g/mol): 116.98 MDL Number: MFCD00011430 InChI Key: YBVKNHXQSRDWAA-UHFFFAOYSA-M PubChem CID: 516859 SMILES: N.O[V](=O)=O
Vanadium Standard For ICP, 10,000μg/mL in 15% HNO3, SPEX CertiPrep™
CAS: 7803-55-6 Molecular Formula: H4NO3V Molecular Weight (g/mol): 116.98 MDL Number: MFCD00011430 InChI Key: YBVKNHXQSRDWAA-UHFFFAOYSA-M PubChem CID: 516859 SMILES: N.O[V](=O)=O
Ammonium vanadium oxide, ACS, 99.0% min
CAS: 7803-55-6 Molecular Formula: H4NO3V Molecular Weight (g/mol): 116.98 MDL Number: MFCD00011430 InChI Key: YBVKNHXQSRDWAA-UHFFFAOYSA-M Synonym: ammonium metavanadate,ammonium vanadate v,ammonium monovanadate,unii-fl85px638g,vanadate vo31-, ammonium,vanadate vo31-, ammonium 1:1,ammonium vanadium oxide,ammoniummetavanadate,vanadic acid, hvo3 , ammonium salt,ammonium meta-vanadate PubChem CID: 516859 SMILES: N.O[V](=O)=O
Ammonium Metavanadate, Extra Pure, SLR, Fisher Chemical™
CAS: 7803-55-6 Molecular Formula: H4NO3V Molecular Weight (g/mol): 116.98 MDL Number: MFCD00011430 InChI Key: YBVKNHXQSRDWAA-UHFFFAOYSA-M IUPAC Name: vanadiumoylol amine SMILES: N.O[V](=O)=O
Ammonium Metavanadate, puriss. p.a. ACS Reagent, ≥99.0% (RT), Solstice
CAS: 7803-55-6 Molecular Formula: H4NO3V Molecular Weight (g/mol): 116.98 MDL Number: MFCD00011430 InChI Key: YBVKNHXQSRDWAA-UHFFFAOYSA-M Synonym: ammonium metavanadate,ammonium vanadate v,ammonium monovanadate,unii-fl85px638g,vanadate vo31-, ammonium,vanadate vo31-, ammonium 1:1,ammonium vanadium oxide,ammoniummetavanadate,vanadic acid, hvo3 , ammonium salt,ammonium meta-vanadate PubChem CID: 516859 IUPAC Name: azanium;oxido(dioxo)vanadium SMILES: N.O[V](=O)=O
Ammonium Metavanadate, ACS Reagent, ≥99%, Solstice
CAS: 7803-55-6 Molecular Formula: H4NO3V Molecular Weight (g/mol): 116.98 MDL Number: MFCD00011430 InChI Key: YBVKNHXQSRDWAA-UHFFFAOYSA-M Synonym: ammonium metavanadate,ammonium vanadate v,ammonium monovanadate,unii-fl85px638g,vanadate vo31-, ammonium,vanadate vo31-, ammonium 1:1,ammonium vanadium oxide,ammoniummetavanadate,vanadic acid, hvo3 , ammonium salt,ammonium meta-vanadate PubChem CID: 516859 IUPAC Name: azanium;oxido(dioxo)vanadium SMILES: N.O[V](=O)=O
- 1
- 2