missing translation for 'onlineSavingsMsg'
Learn More

Ammonium metavanadate, ACS reagent

Product Code. 12605077 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
12605077 25 g Glass bottle 25g
12615077 100 g Glass bottle 100g
12625077 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12605077 Supplier Thermo Scientific Acros Supplier No. 447260250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7803-55-6 | H4NO3V | 116.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7803-55-6
Molecular Formula H4NO3V
Molecular Weight (g/mol) 116.98
MDL Number MFCD00011430
InChI Key YBVKNHXQSRDWAA-UHFFFAOYSA-M
Synonym ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate
PubChem CID 516859
SMILES N.O[V](=O)=O

Specifications

Melting Point 200.0°C
Color White to Yellow
Physical Form Crystalline Solid or Powder
Quantity 25 g
Linear Formula NH4VO3
Merck Index 15,565
Solubility Information (in Ammonium hydroxide) Passes test
IUPAC Name vanadiumoylol amine
Formula Weight 116.98
Percent Purity ≥99.0% (Titrimetry other)
Grade ACS Reagent
Packaging Glass bottle
Chemical Name or Material Ammonium metavanadate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ammonium metavanadate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
  • H319-Causes serious eye irritation.
  • H361d-Suspected of damaging the unborn child.
  • H411-Toxic to aquatic life with long lasting effects.
  • H332-Harmful if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.