missing translation for 'onlineSavingsMsg'
Learn More

Ammonium metavanadate, Puriss. p.a., Reag. Ph. Eur., ≥99.5%, Solstice

Product Code. 15623290 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
1 kg
50 kg
Unit Size:
100g
1kg
50kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15623290 50 kg 50kg
15603290 100 g 100g
15613290 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15623290 Supplier Honeywell-Fluka Supplier No. 3115350KG
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a., Reag. Ph. Eur.,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7803-55-6
Molecular Formula H4NO3V
Molecular Weight (g/mol) 116.98
MDL Number MFCD00011430
InChI Key YBVKNHXQSRDWAA-UHFFFAOYSA-M
Synonym ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate
PubChem CID 516859
IUPAC Name vanadiumoylol amine
SMILES N.O[V](=O)=O

Specifications

CAS Max % 100.0000%
Quantity 50 kg
Linear Formula Ba(OH)2 · 8H2O
UN Number UN2859
Formula Weight 116.98
Percent Purity >99.5%
Grade puriss. p.a.
Packaging Fiberboard Box
Density 2.32 g/cm3 at 25 °C(lit.)
Chemical Name or Material Ammonium metavanadate, Puriss. p.a., Reag. Ph. Eur., ≥99.5%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.