missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Molybdate R2 solution EU Pharmacopoeia, Fisher Chemical™
BIOPHARMA

Product Code. 12940354 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12940354 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12940354 Supplier Fisher Chemical Supplier No. RP 016 1000
  • ONLINE EXCLUSIVE PRICE
    176.00 €
    165.66 € / 1000mL
    Save 10.34 €
    5% Off
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

Packaging Bottle
CAS 12054-85-2, 7732-18-5
Molecular Formula H6MoNO2
Synonym Molybdic acid ammonium salt tetrahydrate, Hexammonium heptamolybdate 4-hydrate
InChI Key XCOMAVCHADVANQ-UHFFFAOYSA-O
SMILES [NH4+].O.[O--].[Mo]
IUPAC Name ammonium hydrate molybdenum oxidandiide
Molecular Weight (g/mol) 148.00
PubChem CID 3084102  
Quantity 1000 mL
Grade EUR Ph Ref: 1005703
Chemical Name or Material Ammonium molybdate solution
Show More Show Less
Product Identifier
  • Ammonium molybdate solution R2. European Pharmacopoeia Ref: 1005703. (RP 016)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.