missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulphate solution 0.5mol/L, Fisher Chemical™

Product Code. 12910914 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12910914 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12910914 Supplier Fisher Chemical Supplier No. VL 204 1000
  • ONLINE EXCLUSIVE PRICE
    198.27 €
    129.17 € / 1000mL
    Save 69.10 €
    34% Off
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Bottle
CAS 7783-20-2
Molecular Formula H8N2O4S
Concentration 0.5 M
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
SMILES N.N.OS(O)(=O)=O
IUPAC Name sulfuric acid diamine
Molecular Weight (g/mol) 132.13
PubChem CID 6097028
Quantity 1000 mL
ChEBI CHEBI:62946
Chemical Name or Material Ammonium Sulphate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.