missing translation for 'onlineSavingsMsg'
Learn More

Ammonium thioglycolate, 70 wt.% solution in water

Product Code. 10268713 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
This item is not returnable. View return policy

Product Code. 10268713

Brand: Thermo Scientific Acros 384682500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ammonium thioglycolate, 68.0 to 72.0%, C2H7NO2S, CAS Number-5421-46-5, 7732-18-5 | CAS: 5421-46-5 | C2H7NO2S | 109.143 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ammonium thioglycolate
Color Colorless
Name Note 70 wt.% solution in water
CAS 5421-46-5, 7732-18-5
Boiling Point 115.0°C
Melting Point -20.0°C
Molecular Formula C2H7NO2S
MDL Number MFCD00137451
Density 1.2200g/mL
Synonym ammonium thioglycolate, ammonium mercaptoacetate, acetic acid, mercapto-, monoammonium salt, thiofaco a-50, ammonium thioglycollate, thioglycolic acid ammonium salt, usaf mo-2, unii-6p9e788vzz, thioglycollic acid, ammonium salt, ammonium 2-mercaptoacetate
Solubility Information Solubility in water: completely soluble.
InChI Key ZZTCCAPMZLDHFM-UHFFFAOYSA-N
SMILES C(C(=O)[O-])S.[NH4+]
IUPAC Name azanium;2-sulfanylacetate
Molecular Weight (g/mol) 109.143
PubChem CID 21534
Formula Weight 109.15
Percent Purity 68.0 to 72.0%
Identification ammonium: positive, mercaptane: positive
Packaging Glass bottle
Specific Gravity 1.22
pH 5.8 to 6.4
Quantity 250 mL
Physical Form Solution
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ammonium thioglycolate, 70 wt.% solution in water
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Substances and mixtures corrosive to metals Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H301-Toxic if swallowed.
  • H317-May cause an allergic skin reaction.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : Ammonium thioglycolate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.