missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amnionless/AMN Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-91203-25ul
This item is not returnable.
View return policy
Description
Amnionless/AMN Polyclonal specifically detects Amnionless/AMN in Human, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA.
Specifications
| Amnionless/AMN | |
| Polyclonal | |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000, Sandwich ELISA | |
| Q9BXJ7 | |
| AMN | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:VSKLWVPNTDFDVAANWSQNRTPCAGGAVEFPADKMVSVLVQEGHAVSDMLLPLDGELVLASGAGFGVSDVGSHLDCGAGEPAVFRDSDRFSWHDPHLWRSGDEAPGLFFVDAERVPCRHDDVFFPPSASFRVGLGPGASPVRVRSISAL | |
| 25 μL | |
| metabolism | |
| 81693 | |
| Human, Rat | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| amnionless homolog (mouse), PRO1028, protein amnionless, UNQ513/PRO1028, visceral endoderm-specific type 1 transmembrane protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of human Amnionless/AMN antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction