missing translation for 'onlineSavingsMsg'
Learn More

Amodiaquine dihydrochloride dihydrate, MedChemExpress

Product Code. 16488726
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488726 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16488726 Supplier Medchem Express Supplier No. HYB1322
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Amodiaquine dihydrochloride dihydrate (Amodiaquin dihydrochloride dihydrate), a 4-aminoquinoline class of antimalarial agent, is a potent and orally active histamine N-methyltransferase inhibitor. Amodiaquine dihydrochloride dihydrate is also a Nurr1 agonist and specifically binds to Nurr1-LBD (ligand binding domain) with an EC50 of ∼20 μM. Anti-inflammatory effect.

Amodiaquine dihydrochloride dihydrate (Amodiaquin dihydrochloride dihydrate), a 4-aminoquinoline class of antimalarial agent, is a potent and orally active histamine N-methyltransferase inhibitor. Amodiaquine dihydrochloride dihydrate is also a Nurr1 agonist and specifically binds to Nurr1-LBD (ligand binding domain) with an EC50 of ∼20 μM. Anti-inflammatory effect.

Specifications

Chemical Name or Material Amodiaquine dihydrochloride dihydrate
CAS 6398-98-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H22ClN3O.2H2O.2HCl
Quantity 100 mg
Synonym Amodiaquin dihydrochloride dihydrate
Solubility Information DMSO : 100 mg/mL (215.14 mM; Need ultrasonic) ∣H2O : 20 mg/mL (43.03 mM; Need ultrasonic)
SMILES OC1=CC=C(NC2=CC=NC3=CC(Cl)=CC=C23)C=C1CN(CC)CC.Cl.Cl.O.O
Molecular Weight (g/mol) 464.81
Formula Weight 464.81
Percent Purity 99.73%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.