missing translation for 'onlineSavingsMsg'
Learn More

Amorolfine hydrochloride, MedChemExpress

Product Code. 16448186
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
Unit Size:
100mg
200mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16448186 100 mg 100mg
16401406 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16448186 Supplier Medchem Express Supplier No. HYB0238
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Amorolfine hydrochloride (Ro 14-4767/002) is a antifungal reagent.

Amorolfine hydrochloride (Ro 14-4767/002) is a antifungal reagent.

Specifications

Chemical Name or Material Amorolfine hydrochloride
Physical Form Solid
CAS 78613-38-4
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C21H36ClNO
For Use With (Application) COVID-19-immunoregulation
Synonym Ro 14-4767/002
Solubility Information DMSO : 12.5 mg/mL (35.31 mM; Need ultrasonic) ∣H2O : 3.33 mg/mL (9.41 mM; Need ultrasonic)
SMILES C[C@@H]1O[C@H](C)CN(CC(C)CC2=CC=C(C(C)(C)CC)C=C2)C1.Cl
Molecular Weight (g/mol) 353.97
Formula Weight 353.97
Percent Purity 99.69%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.