missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMOT Antibody (2C9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42926
This item is not returnable.
View return policy
Description
AMOT Monoclonal antibody specifically detects AMOT in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| AMOT | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 154796 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2b |
| ELISA, Immunohistochemistry, Western Blot | |
| 2C9 | |
| Western Blot 1:1000 - 1:10000, ELISA, Immunohistochemistry 1:20 - 1:200 | |
| angiomotin, angiomotin p80 isoform, KIAA1071angiomotin p130 isoform | |
| angiomotin - (Uniprot# Q4VCS5) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction