missing translation for 'onlineSavingsMsg'
Learn More

Amoxapine, MedChemExpress

Product Code. 16459875
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16459875 250 mg 250mg
16442236 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16459875 Supplier Medchem Express Supplier No. HYB0991
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Amoxapine is a tetracyclic antidepressant of the dibenzoxazepine family, though it is often classified as a secondary amine tricyclic antidepressant.

Amoxapine is a tetracyclic antidepressant of the dibenzoxazepine family, though it is often classified as a secondary amine tricyclic antidepressant.

Specifications

Chemical Name or Material Amoxapine
CAS 14028-44-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C17H16ClN3O
Quantity 250 mg
Synonym CL-67772
Solubility Information DMSO : 8.33 mg/mL (26.55 mM; Need ultrasonic)
SMILES ClC1=CC=C(OC2=CC=CC=C2N=C3N4CCNCC4)C3=C1
Molecular Weight (g/mol) 313.78
Formula Weight 313.78
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.