missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Amoxicillin, 96%

Product Code. 15384988 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15384988 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15384988 Supplier Thermo Scientific Chemicals Supplier No. 455140250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 26787-78-0 | C16H19N3O5S | 365.404 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Type Amoxicillin
Physical Form Solid
CAS 26787-78-0
Melting Point 330.0°C
Odor Odorless
Quantity 25 g
Molecular Formula C16H19N3O5S
MDL Number MFCD00056860
InChI Key LSQZJLSUYDQPKJ-NJBDSQKTSA-N
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=C(C=C3)O)N)C(=O)O)C
IUPAC Name (2S,5R,6R)-6-[[(2R)-2-amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Molecular Weight (g/mol) 365.404
PubChem CID 33613
ChEBI CHEBI:2676
Formula Weight 365.4
Percent Purity 96%
Concentration or Composition (by Analyte or Components) 4-Thia-1-azobicyclo[3.2.0]heptane-2-carboxylic acid,6-[[amino(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-,[2S-[2.alpha.,5.alpha.,6.beta.(S)]]-
Chemical Name or Material Amoxicillin
Show More Show Less
compliance-icons
Product Identifier
  • Amoxicillin
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.