missing translation for 'onlineSavingsMsg'
Learn More

Amoxicillin Crystalline MP Biomedicals

Product Code. 17620897 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Unit Size:
10g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17620897 25 g 25g
17690887 1 g 1g
17600897 5 g 5g
17610897 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17620897 Supplier MP Biomedicals Supplier No. 0219014525
  • ONLINE EXCLUSIVE PRICE
    412.00 €
    390.60 € / 25g
    Save 21.40 €
    5% Off
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 26787-78-0 | Formula: C16H19N3O5S | Weight: 365.404 g/mol

Chemical Identifiers

CAS 26787-78-0
Molecular Formula C16H19N3O5S
Molecular Weight (g/mol) 365.404
InChI Key LSQZJLSUYDQPKJ-NJBDSQKTSA-N
Synonym Amoxycillin, Amolin, AMPC
PubChem CID 33613
ChEBI CHEBI:2676
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=C(C=C3)O)N)C(=O)O)C

Specifications

Type Amoxicillin
Physical Form Crystalline
Quantity 25 g
IUPAC Name (2S,5R,6R)-6-[[(2R)-2-amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Formula Weight 365.4
Percent Purity 95.0 to 102.0%
Grade Not Available
Chemical Name or Material Amoxicillin
compliance-icons compliance-icons
Product Identifier
  • Amoxicillin
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Amoxicillin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.