missing translation for 'onlineSavingsMsg'
Learn More

Amoxicillin, MedChemExpress

Product Code. 16458498
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458498 10 g 10g
16452189 1 g 1g
16440948 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16458498 Supplier Medchem Express Supplier No. HYB0467A
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Amoxicillin (Amoxycillin) is an antibiotic with good oral absorption and broad spectrum antimicrobial activity. Amoxicillin inhibits the biosynthesis of polypeptides in the cell wall, thereby inhibiting cell growth.

Amoxicillin (Amoxycillin) is an antibiotic with good oral absorption and broad spectrum antimicrobial activity. Amoxicillin inhibits the biosynthesis of polypeptides in the cell wall, thereby inhibiting cell growth.

Specifications

Chemical Name or Material Amoxicillin
Physical Form Powder
CAS 26787-78-0
Color White
Quantity 10 g
Purity Grade Notes Research
Molecular Formula C16H19N3O5S
For Use With (Application) COVID-19-immunoregulation
Synonym Amoxycillin
Solubility Information DMSO : 83.33 mg/mL (228.05 mM; Need ultrasonic) ∣H2O : 2 mg/mL (5.47 mM; Need ultrasonic)
SMILES OC([C@@H]1N(C2=O)[C@]([C@@H]2NC([C@@H](C3=CC=C(O)C=C3)N)=O)([H])SC1(C)C)=O
Molecular Weight (g/mol) 365.4
Formula Weight 365.4
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.