Learn More
Melford Amoxicillin Trihydrate
Description
Amoxicillin is a broad-spectrum, semi-synthetic penicillin antibiotic that is effective against a wide range of bacteria.
Amoxicillin is a broad-spectrum, semi-synthetic penicillin antibiotic that is effective against a wide range of bacteria. The trihydrate form of amoxicillin is a crystalline powder that is often used in laboratory settings.
Amoxicillin inhibits the growth of bacteria by interfering with their cell wall synthesis. It is a versatile antibiotic with low toxicity to mammalian cells, making it a valuable tool in maintaining sterile conditions in cell cultures and studying bacterial physiology.
Researchers use amoxicillin to selectively culture bacteria or to treat bacterial infections in cell cultures.
Specifications
Specifications
| Chemical Name or Material | Amoxicillin Trihydrate |
| CAS | 61336-70-7 |
| Quantity | 10 g |
| Molecular Formula | C16H19N3O5S - 3H2O |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.