missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMPD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
226.00 € - 372.00 €
Specifications
| Antigen | AMPD1 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 0.5-2 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18100512
|
Novus Biologicals
NBP2-24509SS |
0.025 mg |
226.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18729273
|
Novus Biologicals
NBP2-24509 |
0.1 mg |
372.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AMPD1 Polyclonal specifically detects AMPD1 in Human, Mouse, Rat, Bovine, Canine, Opossum, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| AMPD1 | |
| Western Blot 0.5-2 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat, Bovine, Canine, Opossum, Primate | |
| adenosine monophosphate deaminase 1, adenosine monophosphate deaminase 1 (isoform M), adenosine monophosphate deaminase-1 (muscle), AMP deaminase 1, AMP deaminase isoform M, AMPD, EC 3.5.4.6, MAD, MADA, Myoadenylate deaminase, skeletal muscle AMPD | |
| AMPD1 | |
| IgG | |
| Affinity Purified | |
| 100% homologous in human (isoforms CRA_a and CRA_c), mouse, rat (isoforms CRA_a and CRA_b). |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 270 | |
| A portion of amino acids 140-190 of human AMPDA1 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title