missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Amphotericin B, 85%

Product Code. 15535156 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15535156 1 g 1g
15545156 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15535156 Supplier Thermo Scientific Chemicals Supplier No. 455490010
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Amphotericin B, CAS # 1397-89-3, is an antifungal that has been used against fungus caused by protozoan parasites of Leishmania genus. | CAS: 1397-89-3 | C47H73NO17 | 924.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Amphotericin B is an antibiotic that has antifungal activity and is produced mainly by Streptomyces nodosus

Applications

• It has antifungal activity against fungus caused by protozoan parasites of Leishmania genus by binding to an essential component of the fungal cell membrane and altering cell membrane permeability

• It can suppress bone marrow progenitor cells and induce Actinomycin D(CR001) sensitivity in drug resistant HELA cells in vitro

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1397-89-3
Molecular Formula C47H73NO17
Molecular Weight (g/mol) 924.09
MDL Number MFCD00877763
InChI Key APKFDSVGJQXUKY-ZNVUZQDLSA-N
PubChem CID 134129663
SMILES C[C@H]1O[C@@H](O[C@@H]2C[C@@H]3O[C@@](O)(C[C@H](O)[C@H]3C(O)=O)C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)\C=C/C=C\C=C/C=C\C=C/C=C\C=C/2)[C@@H](O)[C@@H](N)[C@@H]1O

Specifications

Type Amphotericin B
Melting Point >170.0°C
Water 10% max.
Quantity 1 g
Solubility Information Solubility in water: insoluble.
IUPAC Name (1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19Z,21Z,23Z,25Z,27Z,29Z,31Z,33R,35S,36R,37S)-33-{[(2R,3S,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy}-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid
Formula Weight 924.09
Percent Purity 85%
Chemical Name or Material Amphotericin B
compliance-icons
Product Identifier
  • Amphotericin B
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.