missing translation for 'onlineSavingsMsg'
Learn More

Amphotericin B, MedChemExpress

Product Code. 16488176
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488176 100 mg 100mg
16422099 1 g 1g
16452066 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16488176 Supplier Medchem Express Supplier No. HYB0221
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Amphotericin B is a polyene antifungal agent against a wide variety of fungal pathogens. It binds irreversibly to ergosterol, resulting in disruption of membrane integrity and ultimately cell death.

Amphotericin B is a polyene antifungal agent against a wide variety of fungal pathogens. It binds irreversibly to ergosterol, resulting in disruption of membrane integrity and ultimately cell death.

Specifications

Chemical Name or Material Amphotericin B
Physical Form Powder
CAS 1397-89-3
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C47H73NO17
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 50 mg/mL (54.11 mM; Need ultrasonic)
SMILES C[C@H]([C@@H](O)[C@@H](C)[C@H](C)OC1=O)/C=C/C=C/C=C/C=C/C=C/C=C/C=C/[C@H](O[C@]2([H])O[C@H](C)[C@@H](O)[C@H](N)[C@@H]2O)C[C@@]3([H])[C@H](C(O)=O)[C@@H](O)C[C@](C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)C1)(O)O3
Molecular Weight (g/mol) 924.08
Formula Weight 924.08
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.