missing translation for 'onlineSavingsMsg'
Learn More

Amphotericin B, MP Biomedicals

Product Code. 11420892 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
1 g
5 g
Unit Size:
100mg
1g
250mg
500mg
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420892 250 mg 250mg
17660917 100 mg 100mg
11430892 500 mg 500mg
11440892 1 g 1g
11450892 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11420892 Lieferant MP Biomedicals Lieferanten-Nr. 0219504383
  • ONLINE EXCLUSIVE PRICE
    199.00 €
    187.95 € / 250mg
    Save 11.05 €
    5% Off
Alle Werbeaktionen anzeigen
 Restricted Item
Estimated Shipment: 12-01-2027
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 1397-89-3 | Formula: C47H73NO17 | Weight: 924.09 g/mol

Chemische Identifikatoren

CAS 1397-89-3 , 7732-18-5
Molecular Formula C47H73NO17
Molecular Weight (g/mol) 924.09
MDL Number MFCD00877763
InChI Key APKFDSVGJQXUKY-ZNVUZQDLSA-N
Synonym Amphomoronal, Fungizone, Amphotericine B
PubChem CID 134129663
SMILES C[C@H]1O[C@@H](O[C@@H]2C[C@@H]3O[C@@](O)(C[C@H](O)[C@H]3C(O)=O)C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)\C=C/C=C\C=C/C=C\C=C/C=C\C=C/2)[C@@H](O)[C@@H](N)[C@@H]1O

Spezifikation

Type Amphotericin B
Physical Form Powder
Color Yellow
Quantity 250 mg
IUPAC Name (1S,3R,18S,19R,20R,21S,25R,27R,30R,31R,33S,35R,37S,38R)-3-[(2R,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy-19,25,27,30,31,33,35,37-octahydroxy-18,20,21-trimethyl-23-oxo-22,39-dioxabicyclo[33.3.1]nonatriaconta-4,6,8,10,12,14,16-heptaene-38-car
Formula Weight 924.1
Grade Not Available
Chemical Name or Material Amphotericin B, from Streptomyces sp.
compliance-icons
Produktidentifikator
  • Amphotericin B
Signalwort
  • Warning
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Sicherheitshinweis
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Ergänzende Informationen
  • MIXTURE LIST-Contain: Amphotericin B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.