Learn More
Melford Amphotericin B Solution
Beskrivelse
Amphotericin B 250ug/ml Solution is often utilized in microbiology and cell culture applications.
Amphotericin B 250 μg/mL Solution is often utilized in microbiology and cell culture applications. It is added to growth media to prevent fungal contamination and maintain aseptic conditions in cell cultures.
Fungal cell membranes contain a sterol component called ergosterol, which plays a role in maintaining the integrity and structure of the membrane. Amphotericin B has a high affinity for ergosterol, and it forms complexes with this sterol in the fungal cell membrane.
Once bound to ergosterol, amphotericin B forms channels or pores in the fungal cell membrane. These channels lead to an increase in the permeability of the membrane to ions, particularly potassium, and other molecules. The disruption of the fungal cell membrane compromises its integrity and disrupts the normal functions of the membrane.
The extensive damage to the fungal cell membrane and the loss of cellular components ultimately result in the death of the fungal cell. The antifungal activity of amphotericin B is fungicidal, meaning it directly kills the fungal cells rather than inhibiting their growth.
Tekniske data
Tekniske data
| Chemical Name or Material | Amphotericin B Solution |
| CAS | 1397-89-3 |
| Quantity | 20 mL |
| Molecular Formula | C47H73NO17 |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.