missing translation for 'onlineSavingsMsg'
Learn More

Amphotericin B (Yellow Powder), Fisher BioReagents

Product Code. 10477963
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity unitSize
10477963 250 mg 250mg
1 options
This item is not returnable. View return policy

Product Code. 10477963

Brand: Fisher BioReagents BP928250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
  • Amphotericin B is an antibiotic used for controlling microbial contamination in tissue culture applications
  • Amphotericin B acts by increasing fungal plasma membrane permeability which ultimately leads to fungal cell death
  • Effective in concentration ranges from 0.25 to 2.50 μg/mL
  • Intended for research use only
TRUSTED_SUSTAINABILITY

Specifications

Type Amphotericin B
Physical Form Solid
CAS 1397-89-3
Melting Point 170°C
Ignition Residue < 0.5%
Loss on Drying < 5.0%
Packaging Amber Glass
Potency > 900μg/mg
Color Yellow
Quantity 250 mg
Molecular Formula C47H73NO17
MDL Number MFCD00877763
Merck Index 15, 582
InChI Key APKFDSVGJQXUKY-ZNVUZQDLSA-N
SMILES C[C@H]1O[C@@H](O[C@@H]2C[C@@H]3O[C@@](O)(C[C@H](O)[C@H]3C(O)=O)C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)\C=C/C=C\C=C/C=C\C=C/C=C\C=C/2)[C@@H](O)[C@@H](N)[C@@H]1O
IUPAC Name (1S,3R,18S,19R,20R,21S,25R,27R,30R,31R,33S,35R,37S,38R)-3-[(2R,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy-19,25,27,30,31,33,35,37-octahydroxy-18,20,21-trimethyl-23-oxo-22,39-dioxabicyclo[33.3.1]nonatriaconta-4,6,8,10,12,14,16-heptaene-38-car
Molecular Weight (g/mol) 924.09
PubChem CID 134129663
Formula Weight 924.09
Chemical Name or Material Amphotericin B
Show More Show Less
compliance-icons
Product Identifier
  • Amphotericin B
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.