missing translation for 'onlineSavingsMsg'
Learn More

Ampicillin, MedChemExpress

Product Code. 16482209
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16482209 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16482209 Supplier Medchem Express Supplier No. HYB0522
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Ampicillin is a broad-spectrum beta-lactam antibiotic against a variety of gram-positive and gram-negative bacteria.

Ampicillin is a broad-spectrum beta-lactam antibiotic against a variety of gram-positive and gram-negative bacteria.

Specifications

Chemical Name or Material Ampicillin
Physical Form Powder
CAS 69-53-4
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C16H19N3O4S
For Use With (Application) COVID-19-immunoregulation
Synonym D-(-)-α-Aminobenzylpenicillin
Solubility Information 0.1 M NaOH : 25 mg/mL (71.55 mM; Need ultrasonic) ∣H2O : 5 mg/mL (14.31 mM; ultrasonic and warming and heat to 60°C)
SMILES OC([C@@H]1N(C2=O)[C@]([C@@H]2NC([C@@H](C3=CC=CC=C3)N)=O)([H])SC1(C)C)=O
Molecular Weight (g/mol) 349.4
Formula Weight 349.4
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.