missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Ampicillin ReadyMade™ Solution

Product Code. 30044039
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
5 x 10 mL
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30044039 10 mL 10mL
30044036 5 x 10 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30044039 Supplier TOKU-E™ Supplier No. A22810ML
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Ampicillin ReadyMade™ Solution is provided as a filter-sterilized solution of ampicillin anhydrous formulated in DMSO.

  • CAS Number: 69-53-4
  • Molecular Formula: C16H19N3O4S (ampicillin anhydrous)
  • Molecular Weight: 349.40 g/mol
  • Storage Conditions: -20°C
  • Form: Liquid
  • Source: Semi-synthetic
  • Spectrum: Ampicillin is broad-spectrum, effective against both Gram-positive and Gram-negative bacteria including Gram-negative non-extended spectrum β-lactamase (ESBL) bacteria including Staphylococcus and Streptococcus species and medically important enteric pathogens such as Shigella and Salmonella. Interestingly, Ampicillin has been found to be effective against certain β-lactam sensitive vancomycin-resistant Enterococcus (VRE); a glycopeptide antibiotic resistant 'superbug'. Resistance to Ampicillin is routinely used as a selectable marker to confirm successful cell transformation
  • Mechanism of Action: Like all β-lactams, Ampicillin interferes with penicillin binding protein (PBP) activity, otherwise involved in the final phase of peptidoglycan synthesis. PBPs are enzymes which catalyze a pentaglycine crosslink between alanine and lysine residues. Without a pentaglycine crosslink, the integrity of the cell wall is severely compromised ultimately leading to cell lysis

Specifications

CAS 69-53-4
Certifications/Compliance Purity >98.0%
Chemical Name or Material Ampicillin
Color Opaque white to opaque yellow
Content And Storage -20°C
Description Ampicillin Readymade™ Solution 100 mg/mL in DMSO, Sterile Filtered
Concentration 100 to 105 mg/mL
Form Liquid
Product Type Antibiotic
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives None
Without Additives None
Formulation Liquid, in Dimethyl Sulfoxide
For Use With (Application) Microbiology, Eukaryotic Cell Culture Applications
Product Line ReadyMade™ Solutions
Purity or Quality Grade >98.0%
Quantity 10 mL
Shipping Condition Ice Pack
Source Semi-synthetic
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.