missing translation for 'onlineSavingsMsg'
Learn More

Melford Ampicillin Sodium Salt

Product Code. 30018039
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
1 kg
Unit Size:
100g
10g
1g
1kg
25g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018039 25 g 25g
30018036 10 g 10g
30018037 100 g 100g
30018038 1 kg 1kg
30018040 5 g 5g
30018035 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 30018039 Supplier Melford Supplier No. A4004025.0
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ampicillin, Sodium Salt is an antibiotic that belongs to the penicillin class of antibiotics and is effective against a broad range of Gram-positive and Gram-negative bacteria.

Ampicillin, Sodium Salt is an antibiotic that belongs to the penicillin class of antibiotics and is effective against a broad range of Gram-positive and Gram-negative bacteria. Ampicillin is a semi-synthetic derivative of penicillin and is known for its stability and efficacy.

In the field of molecular biology and genetic engineering, Ampicillin, Sodium Salt is commonly used as a selective agent in plasmid-based cloning experiments. It functions by inhibiting bacterial cell wall synthesis, leading to cell death.

Ampicillin, Sodium Salt is also used in microbiology and cell culture applications. It is effective in preventing the growth of bacterial contaminants in cell cultures, ensuring the purity of cell lines. Additionally, it is employed for the isolation and maintenance of strains with known or engineered resistance to Ampicillin.

One advantage of Ampicillin, Sodium Salt is its stability in aqueous solutions, making it suitable for long-term storage and easy handling. It is available in powdered form and can be easily dissolved in water or other appropriate solvents for use in various applications.

Specifications

Chemical Name or Material Ampicillin, Sodium Salt
CAS 69-52-3
Quantity 25 g
Molecular Formula C16H18N3NaO4S
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.