missing translation for 'onlineSavingsMsg'
Learn More

Ampicillin Trihydrate, MP Biomedicals™

Product Code. 11484582 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484582 25 g 25g
11474582 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11484582 Supplier MP Biomedicals Supplier No. 219014725
  • ONLINE EXCLUSIVE PRICE
    146.00 €
    137.55 € / 25g
    Save 8.45 €
    5% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Active as a broad-spectrum antibiotic. Ampicillin Trihydrate, MP Biomedicals is a semi-synthetic derivative of penicillin that is commonly used to select for ampicillin resistance in mutated and transformed cells. It is also used for enhancing luminol che

Ampicillin trihydrate is a β-lactam antibiotic with an amino group side chain attached to the penicillin structure. It is a penicillin derivative that inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking) by inactivating transpeptidases on the inner surface of the bacterial cell membrane.

  • Inactivated by beta-lactamases; therefore, a beta-lactamase inhibitor should be considered when using ampicillin
  • Antimicrobial spectrum: gram-negative and gram-positive bacteria.
    • Against gram-positive bacteria, ampicillin has a similar mode of action as benzylpenicillin
    • Against gram-negative bacteria, it has a similar mode of action as chloramphenicol and tetracyclines
  • Bactericidal only to growing E. coli. Inhibits cell wall synthesis via cleavage of β-lactam ring of ampicillin by β-lactamase
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7177-48-2
Molecular Formula C16H25N3O7S
Molecular Weight (g/mol) 403.45
InChI Key RXDALBZNGVATNY-CWLIKTDRSA-N
Synonym Aminobenzylpenicillin trihydrate, alpha-Aminobenzylpenicillin trihydrate ;D[-]-α-Aminobenzylpenicillin, 6-[D(-)-α-Aminophenylacetamido]penicillanic acid
PubChem CID 23565
ChEBI CHEBI:31209
IUPAC Name (2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;trihydrate
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C.O.O.O

Specifications

Physical Form Powder
Melting Point 198°C to 200°C (literature)
Concentration Working Concentration: 100mg/liter for both gram positive and gram negative bacteria. It is typically stable in media at 37°C for approximately 3 days.
Quantity 25 g
Solubility Information Soluble in 1M Ammonium Hydroxide (50mg/mL), dilute acids (50mg/mL 1M HCl) or bases; slightly soluble in water (10mg/mL); practically insoluble in alcohol, chloroform, ether and fixed oils.
Formula Weight 403.46
compliance-icons compliance-icons
Product Identifier
  • Ampicillin Trihydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Ampicillin Trihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.