missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ampicillin Trihydrate (Off-white Powder), Fisher BioReagents
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Type | Ampicillin Trihydrate |
| Physical Form | Solid |
| CAS | 7177-48-2 |
| pH | 3.5 to 6.0 |
| Melting Point | 197.8°C |
| Packaging | Poly Bottle |
| Potency | Inclusive between 950μg/mg |
| Color | White |
| Quantity | 25 g |
| Molecular Formula | C16H25N3O7S |
| Show More |
Safety and Handling
Product Identifier
- Ampicillin Trihydrate
- Danger
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction