missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human AMPK alpha-2/beta-1/gamma-3, GST Tag, His Tag Recombinant Protein
Human AMPK alpha-2/beta-1/gamma-3, GST Tag, His Tag Recombinant Protein
Marque: Invitrogen™ A30487
Informations supplémentaires : Weight : 0.10000kg
Description
- This AMPK (A2/B1/G3) recombinant human protein (full length) was expressed in insect cells
- AMPK (AMP-activated protein kinase) serine/threonine kinase is a heterotrimeric complex, which acts as sensor of cellular energy levels
- The signaling cascades initiated by activating AMPK are critical to regulation of metabolic events in the liver, skeletal muscle, heart, adipose tissue, and pancreas
- This AMPK consists of the alpha2, beta1, and gamma3 subunits
- The purified AMPK (A2/B1/G3) complex can be used in a number of different applications
- It has been specifically validated for use with our Z'-LYTE™ assay technology to identify novel kinase inhibitors
- AMPK (A2/B1/G3) recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service
- Validated for use with the Z’-LYTE™ assay
- Available in multiple pack sizes for convenient use
Spécification
PPKAA2: NP_006243, PPKAB1: NP_006244, PPKAG3: NP_059127 | |
89.4/57.5/81.4 kDa | |
CAMKL Kinase Family | |
Human | |
GST Tag, His Tag | |
Dry Ice | |
Kinases & Inhibitors | |
Liquid |
Kinase Assay | |
10 μg | |
Full Length | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol. | |
Baculovirus | |
Full Length, Recombinant, Active |