missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMPK alpha 1 Antibody (2B7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 9 publications
Brand: Novus Biologicals NBP2-22127
This item is not returnable.
View return policy
Description
AMPK alpha 1 Monoclonal specifically detects AMPK alpha 1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| AMPK alpha 1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1 - 2 μg/mL, Simple Western 1:50, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 2 - 4 μg/mL, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| AMP -activate kinase alpha 1 subunit, AMP-activated protein kinase, catalytic, alpha-1,5'-AMP-activated protein kinase catalytic subunit alpha-1, AMPK, AMPK alpha 1,5'-AMP-activated protein kinase, catalytic alpha-1 chain, AMPK subunit alpha-1, AMPK1, AMPKa1, EC 2.7.11, EC 2.7.11.1, MGC33776, MGC57364, protein kinase, AMP-activated, alpha 1 catalytic subunit | |
| Mouse | |
| 64 kDa | |
| 0.1 mL | |
| Apoptosis, Autophagy, Cancer, Hypoxia, MAP Kinase Signaling, mTOR Pathway, Signal Transduction, Translation Control | |
| 5562 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 2B7 | |
| Unconjugated | |
| Q13131 | |
| PRKAA1 | |
| Purified recombinant fragment of human AMPK alpha 1 expressed in E. coli. [UniProt# Q13131] | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Primate | |
| Precipitation |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction