missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMPK alpha 1, p Ser496 Antibody (S01-3B7), Novus Biologicals™
Rabbit Monoclonal Antibody
421.00 € - 645.00 €
Specifications
| Antigen | AMPK alpha 1 |
|---|---|
| Clone | S01-3B7 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18625903
|
Novus Biologicals
NBP3-19825-50ul |
50 μg |
421.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18678373
|
Novus Biologicals
NBP3-19825-100ul |
100 μg |
645.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AMPK alpha 1 Monoclonal antibody specifically detects AMPK alpha 1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| AMPK alpha 1 | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| AMP -activate kinase alpha 1 subunit, AMP-activated protein kinase, catalytic, alpha-1,5'-AMP-activated protein kinase catalytic subunit alpha-1, AMPK, AMPK alpha 1,5'-AMP-activated protein kinase, catalytic alpha-1 chain, AMPK subunit alpha-1, AMPK1, AMPKa1, EC 2.7.11, EC 2.7.11.1, MGC33776, MGC57364, protein kinase, AMP-activated, alpha 1 catalytic subunit | |
| A synthetic phosphopeptide corresponding to residues surrounding Ser496 of human AMPK alpha 1 (Uniprot # Q13131) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| S01-3B7 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Autophagy, Cancer, Hypoxia, MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Signal Transduction, Translation Control | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 5562 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title