missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMPK beta 1 (PRKAB1) Monoclonal Antibody (OTI3B11), OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813115S
This item is not returnable.
View return policy
Description
AMPK beta 1 (PRKAB1) Monoclonal antibody specifically detects AMPK beta 1 (PRKAB1) in Human samples. It is validated for Western BlotSpecifications
| AMPK beta 1 (PRKAB1) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| A6NNA5 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 644168 | |
| -20°C | |
| IgG1 |
| Western Blot | |
| OTI3B11 | |
| Unconjugated | |
| DRGX | |
| 1300015D22Rik; 5'-AMP-activated protein kinase 40 kDa subunit; 5-AMP-activated protein kinase beta subunit; 5'-AMP-activated protein kinase beta-1 subunit; 5'-AMP-activated protein kinase subunit beta-1; 5'-AMP-activated protein kinase subunit beta-1-like protein; 5'-AMP-activated protein kinase, beta subunit; AAKB1; AMP-activated protein kinase beta subunit; AMPK; AMPK beta 1; AMPK beta -1 chain; AMPK beta-1 chain; AMPK subunit beta-1; AMPKb; AMPK-beta1; AU021155; E430008F22; HAMPKb; kinase AMPK-beta1; kinase pAMPK; pAMPK; Prkab1; protein kinase AMP-activated non-catalytic subunit beta 1; protein kinase, AMP-activated, beta 1 non-catalytic subunit; protein kinase, AMP-activated, noncatalytic, beta-1 | |
| Human recombinant protein fragment corresponding to amino acids 1-263 of human DRGX (NP_001073989) produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction