missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AMPK gamma-1 Recombinant Rabbit Monoclonal Antibody (24GB3285)

Rabbit Recombinant Monoclonal Antibody
146.00 € - 500.00 €
Specifications
| Antigen | AMPK gamma-1 |
|---|---|
| Clone | 24GB3285 |
| Content And Storage | -20°C |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275277
![]() |
Invitrogen™
MA557187 |
100 μL |
500.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275050
|
Invitrogen™
MA557188 |
20 μL |
146.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AMPK gamma-1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
The protein encoded by this gene is a regulatory subunit of the AMP-activated protein kinase (AMPK). AMPK is a heterotrimer consisting of an alpha catalytic subunit, and non-catalytic beta and gamma subunits. AMPK is an important energy-sensing enzyme that monitors cellular energy status. In response to cellular metabolic stresses, AMPK is activated, and thus phosphorylates and inactivates acetyl-CoA carboxylase (ACC) and beta-hydroxy beta-methylglutaryl-CoA reductase (HMGCR), key enzymes involved in regulating de novo biosynthesis of fatty acid and cholesterol. This subunit is one of the gamma regulatory subunits of AMPK. Alternatively spliced transcript variants encoding distinct isoforms have been observed.Specifications
| AMPK gamma-1 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| PRKAG1 | |
| 5'-AMP-activated protein kinase subunit gamma-1; 5'-AMP-activated protein kinase, gamma-1 subunit; AA571379; AMP-activated protein kinase, noncatalytic gamma-1 subunit; ampk gamma; AMPK gamma1; AMPK gamma-1 chain; AMPK subunit gamma-1; AMPKG; BB036179; Prkaac; PRKAG1; Prkga1; Protein kinase AMP-activated gamma; protein kinase AMP-activated non-catalytic subunit gamma 1; protein kinase, AMP-activated, gamma 1 non-catalytic subunit | |
| PRKAG1 | |
| Primary | |
| Affinity chromatography |
| 24GB3285 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O54950, P54619, P80385 | |
| 19082, 25520, 5571 | |
| A synthesized peptide derived from human AMPK gamma 1 (aa 280-331). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title