missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Anaerobe Blood Agar (Wilkins Chalgren) with Neomycin

Product Code. 12907128
Change view
Click to view available options
Description:
Anaerobe Blood Agar (WC) with Neomycin
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
12907128 Anaerobe Blood Agar (WC) with Neomycin Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12907128 Supplier Thermo Scientific™ Supplier No. PB0112A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate, cultivate and /or carry out susceptibility testing of a wide variety of anaerobic organisms from clinical samples with this selective medium.

Isolate, cultivate and /or carry out susceptibility testing of a wide variety of anaerobic organisms from clinical samples with Thermo Scientific™ Oxoid™ Anaerobe Blood Agar (Wilkins Chalgren) with Neomycin. The addition of Neomycin- an aminoglycoside antibiotic inhibits the growth of most of the facultative Gram-negative anaerobic bacteria while allowing the growth of many other anaerobic bacteria. Anaerobe Blood Agar (Wilkins Chalgren) with Neomycin is a modification of the medium described by Wilkins and Chalgren1.

Use Anaerobe Blood Agar with Neomycin for the isolation of general anaerobes from clinical samples and for antimicrobial susceptibility testing of aerobic bacteria using Receiver Operating Characteristic (ROC) procedure.

  • Selective for certain anaerobic organisms: the presence of Neomycin, an aminoglycoside antibiotic is bactericidal to many Gram negative facultative anaerobes and some staphylococci. This is added to the medium to inhibit accompanying flora.
  • Ready-to-use convenience of prepared media.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Red
Content And Storage 2°C to 10°C
Description Anaerobe Blood Agar (WC) with Neomycin
Format 90mm Monoplate
Manufacturing Origin United Kingdom
Product Type Agar Plate
Fill Weight 19.5g±1.0.g
Packaging Type 10 x 90mm plates, film wrapped
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.