missing translation for 'onlineSavingsMsg'
Learn More

Analytes B For Environmental Analysis, SPEX CertiPrep™

Product Code. 15658305 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
125 mL
Unit Size:
125mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15658305 125 mL 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15658305 Supplier SPEX Certiprep Supplier No. INTB3
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Cd100μg/mL
Ni100μg/mL
Zn100μg/mL
Sb60μg/mL
Ba50μg/mL
Be50μg/mL
Co50μg/mL
Cr50μg/mL
Cu50μg/mL
Mn50μg/mL
V50μg/mL
Ag20μg/mL
As10μg/mL
Tl10μg/mL
Pb5μg/mL
Se5μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Analytes B3
Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Nitric Acid/Trace Tartaric Acid/Trace Hydrogen Fluoride
Quantity 125 mL
Grade Environmental
Concentration or Composition (by Analyte or Components) Components in 5% HNO3/tr Tartaric Acid/tr HF:
Sb    •60μg/mL
As    •10μg/mL
Ba    •50μg/mL
Be    •50μg/mL
Cd    •100μg/mL
Cr    •50μg/mL
Co    •50μg/mL
Cu    •50μg/mL
Pb    •5μg/mL
Mn    •50μg/mL
Ni    •100μg/mL
Se    •5μg/mL
Ag    •20μg/mL
Tl    •10μg/mL
V    •50μg/mL
Zn    •100μg/mL
compliance-icons
Product Identifier
  • Analytes B
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: nitric acid, hydrofluoric acid

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.