missing translation for 'onlineSavingsMsg'
Learn More

Andrographolide, MedChemExpress

Product Code. 16469026
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16469026 100 mg 100mg
16442366 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16469026 Supplier Medchem Express Supplier No. HYN0191
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Andrographolide is a NF-κB inhibitor, which inhibits NF-κB activation through covalent modification of a cysteine residue on p50 in endothelial cells without affecting IκBα degradation or p50/p65 nuclear translocation. Andrographolide has antiviral effects.

Andrographolide is a NF-κB inhibitor, which inhibits NF-κB activation through covalent modification of a cysteine residue on p50 in endothelial cells without affecting IκBα degradation or p50/p65 nuclear translocation. Andrographolide has antiviral effects.

Specifications

Chemical Name or Material Andrographolide
Physical Form Solid
CAS 5508-58-7
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C20H30O5
For Use With (Application) Cancer-programmed cell death
Synonym Andrographis
Solubility Information DMSO : 100 mg/mL (285.35 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C1OC[C@@H](O)/C1=C\C[C@@H]2C(CC[C@]3([H])[C@](C)(CO)[C@H](O)CC[C@@]23C)=C
Molecular Weight (g/mol) 350.45
Formula Weight 350.45
Percent Purity 97.46%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.